Assay Details
| Gene Symbol: Wibg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506200.2 | XP_003506248.2 | 1 - 2 | 396 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wibg| Gene Symbol: Wibg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506200.2 | XP_003506248.2 | 1 - 2 | 396 | 61 | |
Set Membership: |
|