Assay Details
| Gene Symbol: Rnf217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503204.2 | XP_003503252.2 | 5 - 6 | 1019 | 100 | |
| GenBank mRNA | JI887570.1 | - | 2 - 3 | 269 | 100 | |
More Information
Set Membership: |
|

Rnf217
Genomic Map