Assay Details
| Gene Symbol: Zfp91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654606.1 | XP_007652796.1 | 8 - 9 | 738 | 115 | |
| GenBank mRNA | JP055359.1 | - | 8 - 9 | 730 | 114 | |
More Information
Set Membership: |
|

Zfp91
Genomic Map