Assay Details
Gene Symbol: Syngr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498835.2 | XP_003498883.2 | 2 - 3 | 494 | 84 | |
GenBank mRNA | JI886370.1 | - | 2 - 3 | 494 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
