Assay Details
Gene Symbol: Dazl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500899.1 | XP_003500947.1 | 4 - 5 | 339 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
