Assay Details
| Gene Symbol: Hcfc1r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508712.2 | XP_003508760.1 | 2 - 3 | 364 | 102 | |
| XM_007649335.1 | XP_007647525.1 | 2 - 3 | 469 | 102 | ||
| GenBank mRNA | JI869375.1 | - | 1 - 2 | 134 | 102 | |
| JP059050.1 | - | 2 - 3 | 408 | 102 | ||
More Information
Set Membership: |
|

Hcfc1r1
Genomic Map