Assay Details
Gene Symbol: Fzd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497961.2 | XP_003498009.1 | 5 - 6 | 2102 | 84 | |
XM_007620234.1 | XP_007618424.1 | 4 - 5 | 1542 | 84 | ||
GenBank mRNA | JI864998.1 | - | 1 - 2 | 144 | 84 | |
JP039813.1 | - | 1 - 2 | 144 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
