Assay Details
Gene Symbol: Bnip3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007648768.1 | XP_007646958.1 | 3 - 4 | 553 | 84 | |
GenBank mRNA | JI882681.1 | - | 3 - 4 | 556 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
