Assay Details
| Gene Symbol: Fam103a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511808.2 | XP_003511856.1 | 2 - 3 | 561 | 86 | |
| GenBank mRNA | JI884918.1 | - | 2 - 3 | 322 | 86 | |
| JP053340.1 | - | 2 - 3 | 322 | 86 | ||
More Information
Set Membership: |
|

Fam103a1
Genomic Map