Assay Details
Gene Symbol: Hspa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246739.2 | NP_001233668.1 | 8 - 9 | 1554 | 87 | |
GenBank mRNA | JI874710.1 | - | 8 - 9 | 1554 | 87 | |
JP038542.1 | - | 8 - 9 | 1554 | 87 | ||
M17169.1 | - | 8 - 9 | 1555 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
