Assay Details
Gene Symbol: Ndfip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643053.1 | XP_007641243.1 | 7 - 8 | 649 | 82 | |
GenBank mRNA | JP061364.1 | - | 5 - 6 | 558 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
