Assay Details
Gene Symbol: Ammecr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007651423.1 | XP_007649613.1 | 2 - 3 | 561 | 70 | |
GenBank mRNA | JI862746.1 | - | 792 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
