Assay Details
| Gene Symbol: Rhoc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512964.2 | XP_003513012.1 | 1 - 2 | 216 | 93 | |
| GenBank mRNA | JI873603.1 | - | 1 - 2 | 216 | 93 | |
| JP052796.1 | - | 1 - 2 | 216 | 93 | ||
More Information
Set Membership: |
|

Rhoc
Genomic Map