Assay Details
| Gene Symbol: Hm13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642929.1 | XP_007641119.1 | 4 - 5 | 473 | 82 | |
| GenBank mRNA | JI861980.1 | - | 4 - 5 | 309 | 82 | |
More Information
Set Membership: |
|

Hm13
Genomic Map