Assay Details
Gene Symbol: Rab30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507015.2 | XP_003507063.1 | 2 - 3 | 298 | 63 | |
XM_007647545.1 | XP_007645735.1 | 3 - 4 | 435 | 63 | ||
XM_007647546.1 | XP_007645736.1 | 2 - 3 | 150 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
