Assay Details
| Gene Symbol: Ppp2r1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649966.1 | XP_007648156.1 | 5 - 6 | 972 | 92 | |
| GenBank mRNA | JI896774.1 | - | 5 - 6 | 972 | 92 | |
| JP043398.1 | - | 5 - 6 | 972 | 92 | ||
More Information
Set Membership: |
|

Ppp2r1a
Genomic Map