Assay Details
| Gene Symbol: Pgp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501553.2 | XP_003501601.1 | 3 - 4 | 814 | 58 | |
| GenBank mRNA | JI863234.1 | - | 1 - 2 | 123 | 58 | |
More Information
Set Membership: |
|

Pgp
Genomic Map