Assay Details
| Gene Symbol: LOC100758105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510138.2 | XP_003510186.1 | 3 - 4 | 1133 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100758105| Gene Symbol: LOC100758105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510138.2 | XP_003510186.1 | 3 - 4 | 1133 | 64 | |
Set Membership: |
|