Assay Details
| Gene Symbol: Ppp1r11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507410.2 | XP_003507458.1 | 2 - 3 | 357 | 74 | |
| GenBank mRNA | JI869620.1 | - | 2 - 3 | 377 | 74 | |
| JP054252.1 | - | 2 - 3 | 377 | 74 | ||
More Information
Set Membership: |
|

Ppp1r11
Genomic Map