Assay Details
Gene Symbol: Csnk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508482.2 | XP_003508530.1 | 4 - 5 | 396 | 53 | |
GenBank mRNA | JI866568.1 | - | 4 - 5 | 499 | 53 | |
JP059390.1 | - | 4 - 5 | 499 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
