Assay Details
| Gene Symbol: Xiap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246801.1 | NP_001233730.1 | 1 - 2 | 900 | 78 | |
| GenBank mRNA | GQ183932.1 | - | 1 - 2 | 900 | 78 | |
More Information
Set Membership: |
|

Xiap
Genomic Map