Assay Details
Gene Symbol: Eif4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508213.2 | XP_003508261.1 | 8 - 9 | 950 | 60 | |
XM_003508215.2 | XP_003508263.1 | 7 - 8 | 808 | 60 | ||
XM_007648841.1 | XP_007647031.1 | 8 - 9 | 906 | 60 | ||
GenBank mRNA | JI878461.1 | - | 4 - 5 | 558 | 60 | |
JP050156.1 | - | 4 - 5 | 559 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
