Assay Details
Gene Symbol: Mt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244576.1 | NP_001231505.1 | 2 - 3 | 92 | 90 | |
GenBank mRNA | JI877540.1 | - | 2 - 3 | 265 | 90 | |
JP043447.1 | - | 2 - 3 | 265 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
