Assay Details
Gene Symbol: Cenpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246717.1 | NP_001233646.1 | 2 - 3 | 275 | 78 | |
XM_007654978.1 | XP_007653168.1 | 2 - 3 | 355 | 78 | ||
GenBank mRNA | AJ428867.1 | - | 2 - 3 | 275 | 78 | |
JI875773.1 | - | 1 - 2 | 48 | 78 | ||
JP061690.1 | - | 1 - 2 | 48 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
