Assay Details
| Gene Symbol: Gosr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246745.1 | NP_001233674.1 | 2 - 3 | 175 | 96 | |
| GenBank mRNA | JI889453.1 | - | 6 - 7 | 96 | ||
| U49841.1 | - | 2 - 3 | 175 | 96 | ||
More Information
Set Membership: |
|

Gosr1
Genomic Map