Assay Details
Gene Symbol: Tlr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246762.1 | NP_001233691.1 | 1 - 2 | 345 | 68 | |
GenBank mRNA | AF153676.1 | - | 1 - 2 | 345 | 68 | |
JI881679.1 | - | 1 - 2 | 401 | 68 | ||
JP060647.1 | - | 1 - 2 | 401 | 68 |

More Information
Set Membership: |
![]() ![]() |
