Assay Details
Gene Symbol: Rps7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285924.1 | NP_001272853.1 | 3 - 4 | 227 | 53 | |
GenBank mRNA | JI871282.1 | - | 2 - 3 | 351 | 53 | |
JP044871.1 | - | 3 - 4 | 227 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
