Assay Details
Gene Symbol: Kin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291665.1 | NP_001278594.1 | 4 - 5 | 375 | 68 | |
GenBank mRNA | JI893007.1 | - | 4 - 5 | 420 | 68 | |
JP058382.1 | - | 4 - 5 | 420 | 68 | ||
KJ193861.1 | - | 4 - 5 | 375 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
