Assay Details
| Gene Symbol: Apex1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244411.1 | NP_001231340.1 | 4 - 5 | 514 | 62 | |
| XM_007649790.1 | XP_007647980.1 | 3 - 4 | 668 | 62 | ||
| GenBank mRNA | JI883500.1 | - | 3 - 4 | 678 | 62 | |
| JP047836.1 | - | 4 - 5 | 561 | 62 | ||
More Information
Set Membership: |
|

Apex1
Genomic Map