Assay Details
| Gene Symbol: Sat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246689.1 | NP_001233618.1 | 5 - 6 | 343 | 70 | |
| GenBank mRNA | AF281149.1 | - | 5 - 6 | 343 | 70 | |
More Information
Set Membership: |
|

Sat1
Genomic Map