Assay Details
| Gene Symbol: Sdhc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246826.1 | NP_001233755.1 | 3 - 4 | 194 | 65 | |
| XM_007647383.1 | XP_007645573.1 | 3 - 4 | 209 | 65 | ||
| GenBank mRNA | JI877771.1 | - | 3 - 4 | 211 | 65 | |
| JP037986.1 | - | 3 - 4 | 211 | 65 | ||
| U31241.1 | - | 3 - 4 | 194 | 65 | ||
More Information
Set Membership: |
|

Sdhc
Genomic Map