Assay Details
Gene Symbol: Cdk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244027.1 | NP_001230956.1 | 2 - 3 | 310 | 58 | |
GenBank mRNA | AJ223949.1 | - | 3 - 4 | 465 | 58 | |
AJ223952.1 | - | 2 - 3 | 310 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
