Assay Details
Gene Symbol: Rps17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244040.1 | NP_001230969.1 | 2 - 3 | 278 | 85 | |
GenBank mRNA | JI881027.1 | - | 3 - 4 | 299 | 85 | |
JP060743.1 | - | 3 - 4 | 299 | 85 | ||
M13933.1 | - | 2 - 3 | 278 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
