Assay Details
| Gene Symbol: Rps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244043.1 | NP_001230972.1 | 1 - 2 | 192 | 55 | |
| XM_007642121.1 | XP_007640311.1 | 2 - 3 | 264 | 55 | ||
| GenBank mRNA | AY392426.1 | - | 1 - 2 | 192 | 55 | |
| JI872398.1 | - | 1 - 2 | 130 | 55 | ||
| JP053273.1 | - | 1 - 2 | 130 | 55 | ||
More Information
Set Membership: |
|

Rps2
Genomic Map