Assay Details
| Gene Symbol: Cxcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244044.1 | NP_001230973.1 | 3 - 4 | 340 | 83 | |
| GenBank mRNA | AB089673.1 | - | 1 - 2 | 60 | 83 | |
| J03560.1 | - | 3 - 4 | 340 | 83 | ||
More Information
Set Membership: |
|

Cxcl1
Genomic Map