Assay Details
Gene Symbol: Dpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246685.1 | NP_001233614.1 | 465 | 78 | ||
GenBank mRNA | AF121895.1 | - | 465 | 78 | ||
JI882481.1 | - | 1 - 2 | 185 | 78 | ||
JP037743.1 | - | 1 - 2 | 185 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
