Assay Details
| Gene Symbol: Cdkn1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246825.2 | NP_001233754.1 | 2 - 3 | 651 | 84 | |
| GenBank mRNA | U49649.1 | - | 2 - 3 | 651 | 84 | |
More Information
Set Membership: |
|

Cdkn1b
Genomic Map