Assay Details
Gene Symbol: Vimp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256848.1 | NP_001243777.1 | 3 - 4 | 382 | 88 | |
GenBank mRNA | JP047584.1 | - | 3 - 4 | 382 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
