Assay Details
Gene Symbol: Vimp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256848.1 | NP_001243777.1 | 4 - 5 | 470 | 76 | |
GenBank mRNA | JI894185.1 | - | 1 - 2 | 87 | 76 | |
JP047584.1 | - | 4 - 5 | 470 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
