Assay Details
Gene Symbol: Tvp23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281883.1 | NP_001268812.1 | 1 - 2 | 119 | 87 | |
GenBank mRNA | JI881887.1 | - | 1 - 2 | 119 | 87 | |
JP038707.1 | - | 1 - 2 | 119 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
