Assay Details
| Gene Symbol: LOC100760212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291946.1 | NP_001278875.1 | 28 - 29 | 3286 | 84 | |
| XM_003507038.1 | XP_003507086.1 | 28 - 29 | 3298 | 84 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100760212| Gene Symbol: LOC100760212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291946.1 | NP_001278875.1 | 28 - 29 | 3286 | 84 | |
| XM_003507038.1 | XP_003507086.1 | 28 - 29 | 3298 | 84 | ||