Assay Details
| Gene Symbol: Abcb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243989.1 | NP_001230918.1 | 17 - 18 | 2322 | 67 | |
| XM_007646774.1 | XP_007644964.1 | 18 - 19 | 2486 | 67 | ||
| XM_007646775.1 | XP_007644965.1 | 16 - 17 | 2261 | 67 | ||
| XM_007646776.1 | XP_007644966.1 | 18 - 19 | 2486 | 67 | ||
| GenBank mRNA | M17896.1 | - | 4 - 5 | 451 | 67 | |
| M60041.1 | - | 17 - 18 | 2322 | 67 | ||
More Information
Set Membership: |
|

Abcb1b
Genomic Map