Assay Details
| Gene Symbol: Abcb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243989.1 | NP_001230918.1 | 2 - 3 | 124 | 98 | |
| XM_007646774.1 | XP_007644964.1 | 3 - 4 | 288 | 95 | ||
| XM_007646776.1 | XP_007644966.1 | 3 - 4 | 288 | 95 | ||
| XM_007646777.1 | XP_007644967.1 | 3 - 4 | 287 | 95 | ||
| GenBank mRNA | M60041.1 | - | 2 - 3 | 124 | 98 | |
More Information
Set Membership: |
|

Abcb1b
Genomic Map