Assay Details
Gene Symbol: Slc35a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244019.1 | NP_001230948.1 | 1 - 2 | 100 | 68 | |
XM_007645599.1 | XP_007643789.1 | 1 - 2 | 392 | 68 | ||
GenBank mRNA | AF299335.1 | - | 1 - 2 | 100 | 68 | |
FN825776.1 | - | 1 - 2 | 97 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
