Assay Details
Gene Symbol: Icam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246690.1 | NP_001233619.1 | 3 - 4 | 803 | 69 | |
GenBank mRNA | AF308456.1 | - | 3 - 4 | 803 | 69 | |
JI866855.1 | - | 3 - 4 | 724 | 69 | ||
JP055129.1 | - | 3 - 4 | 724 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
