Assay Details
| Gene Symbol: Pgk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246725.1 | NP_001233654.1 | 4 - 5 | 460 | 78 | |
| GenBank mRNA | JI880676.1 | - | 4 - 5 | 540 | 78 | |
| JP046257.1 | - | 4 - 5 | 540 | 78 | ||
| Z37974.1 | - | 4 - 5 | 460 | 78 | ||
More Information
Set Membership: |
|

Pgk1
Genomic Map