Assay Details
| Gene Symbol: Cbx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246787.1 | NP_001233716.1 | 538 | 70 | ||
| GenBank mRNA | AY548739.1 | - | 538 | 70 | ||
| JI876171.1 | - | 3 - 4 | 554 | 70 | ||
| JP059074.1 | - | 3 - 4 | 554 | 70 | ||
More Information
Set Membership: |
|

Cbx1
Genomic Map