Assay Details
| Gene Symbol: Oaz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291906.1 | NP_001278835.1 | 2 - 3 | 385 | 67 | |
| GenBank mRNA | JI879565.1 | - | 3 - 4 | 345 | 67 | |
| JP061177.1 | - | 3 - 4 | 345 | 67 | ||
More Information
Set Membership: |
|

Oaz2
Genomic Map