Assay Details
| Gene Symbol: Slc31a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244117.1 | NP_001231046.1 | 2 - 3 | 228 | 67 | |
| GenBank mRNA | HQ290320.1 | - | 2 - 3 | 228 | 67 | |
| JI864695.1 | - | 1 - 2 | 202 | 67 | ||
| JP063222.1 | - | 1 - 2 | 202 | 67 | ||
More Information
Set Membership: |
|

Slc31a1
Genomic Map