Assay Details
Gene Symbol: Tk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244494.1 | NP_001231423.1 | 6 - 7 | 513 | 69 | |
XM_007633677.1 | XP_007631867.1 | 5 - 6 | 639 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
